Learn More
2-(4-Morpholinyl)ethyl isothiocyanate, 97%, Thermo Scientific Chemicals
Description
2-(4-Morpholinyl)ethyl isothiocyanate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(4-Morpholinyl)ethyl isothiocyanate is used as pharmaceutical intermediates.
Solubility
Hydrolyzes in water.
Notes
Store at 4°C. Store under dry inert gas. Protect from humidity and water. Incompatible with moisture, amines, alcohols and oxidizing agents. Protect from heat. Moisture Sensitive.
Specifications
Specifications
| Density | 1.11 |
| Boiling Point | 162°C to 163°C (10 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.539 |
| Quantity | 2 g |
| UN Number | UN2922 |
| Beilstein | 1210530 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Hydrolyzes in water. |
| Formula Weight | 172.25 |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332-H314-H335 |
| Hazard Statement | GHS H Statement H331-H314-H318-H302-H312 Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S. |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only