missing translation for 'onlineSavingsMsg'
Learn More

[2-(4-Methylperhydro-1,4-diazepin-1-yl)phenyl]methanol, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC60409DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC60409DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

915707-55-0
220.32
IYBDLMFCIKDQDV-UHFFFAOYSA-N
24229618
CN1CCCN(CC1)C1=CC=CC=C1CO
C13H20N2O
MFCD09065000
2-4-methylperhydro-1,4-diazepin-1-yl phenyl methanol, 2-4-methyl-1,4-diazepan-1-yl phenyl methanol, 2-4-methylhomopiperazin-1-yl benzyl alcohol, 2-4-methyl-1,4-diazaperhydroepinyl phenyl methan-1-ol
[2-(4-methyl-1,4-diazepan-1-yl)phenyl]methanol

Specifications

[2-(4-Methylperhydro-1, 4-diazepin-1-yl)phenyl]methanol
96.5
97+%
C13H20N2O
MFCD09065000
IYBDLMFCIKDQDV-UHFFFAOYSA-N
[2-(4-methyl-1,4-diazepan-1-yl)phenyl]methanol
24229618
97%
915707-55-0
100.0
Amber Glass Bottle
5g
2-4-methylperhydro-1,4-diazepin-1-yl phenyl methanol, 2-4-methyl-1,4-diazepan-1-yl phenyl methanol, 2-4-methylhomopiperazin-1-yl benzyl alcohol, 2-4-methyl-1,4-diazaperhydroepinyl phenyl methan-1-ol
CN1CCCN(CC1)C1=CC=CC=C1CO
220.32
220.31
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents