missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methyl-6,7-dihydro-4H-thieno[3,2-c]pyran-4-yl)ethanol, 95%, Thermo Scientific™

Numéro de catalogue. p-3541081
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC61809DA 1g Amber Glass Bottle
CC61809DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC61809DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC61809DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 898289-60-6
Formule moléculaire C10H14O2S
Poids moléculaire (g/mol) 198.28
Numéro MDL MFCD09702404
Clé InChI UTWVZRFJFAVPIU-UHFFFAOYNA-N
Synonyme 2-4-methyl-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl ethanol, 2-4-methyl-6h,7h-thieno 3,2-c pyran-4-yl ethanol, 4h-thieno 3,2-c pyran-4-ethanol,6,7-dihydro-4-methyl, 2-4-methyl-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl ethan-1-ol, 4-2-hydroxyethyl-4-methyl-6,7-dihydro-4h-thieno 3,2-c pyran, 2-4-methyl-6,7-dihydro-4h-thiopheno 3,2-c pyran-4-yl ethan-1-ol
PubChem CID 24229641
Nom de l’IUPAC 2-{4-methyl-4H,6H,7H-thieno[3,2-c]pyran-4-yl}ethan-1-ol
SOURIRES CC1(CCO)OCCC2=C1C=CS2

Spécifications

Nom ou substance chimique 2-(4-Methyl-6, 7-dihydro-4H-thieno[3, 2-c]pyran-4-yl)ethanol
CAS % minimum 94.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 95%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 198.28
Pourcentage de pureté 95%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.