missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methyl-5-thiazolyl)ethyl Hexanoate 98.0+%, TCI America™

Numéro de catalogue. p-7133251
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M24915G 5 g
M249125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M24915G Fournisseur TCI America Code fournisseur M24915G
Il en reste null

Identifiants chimiques

CAS 94159-32-7
Molecular Formula C12H19NO2S
Molecular Weight (g/mol) 241.35
MDL Number MFCD09032914
InChI Key VJULDCZELAIZHC-UHFFFAOYSA-N
Synonym Hexanoic Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester, 5-(2-Hexanoyloxyethyl)-4-methylthiazole
PubChem CID 3023839
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethyl hexanoate
SMILES CCCCCC(=O)OCCC1=C(C)N=CS1

Spécifications

Boiling Point 152°C
Quantity 5 g
Formula Weight 241.35
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Methyl-5-thiazolyl)ethyl Hexanoate