missing translation for 'onlineSavingsMsg'
Learn More

2,4-Lutidine 98.0+%, TCI America™

Catalog No. L0085500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0085500ML 500 mL
L008525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0085500ML Supplier TCI America Supplier No. L0085500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-47-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00006337
InChI Key JYYNAJVZFGKDEQ-UHFFFAOYSA-N
Synonym 2,4-lutidine, pyridine, 2,4-dimethyl, alpha,gamma-dimethylpyridine, 2,4-lutidene, unii-83903uj0ww, 2,4-dimethyl-pyridine, .alpha.,.gamma.-dimethylpyridine, 2,4-dimethyl pyridine, 2.4-lutidine, 2,4 lutidine
PubChem CID 7936
IUPAC Name 2,4-dimethylpyridine
SMILES CC1=CC=NC(C)=C1

Specifications

Melting Point -64°C
Color Yellow
Boiling Point 157°C
Quantity 500 mL
UN Number 1992
Formula Weight 107.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Lutidine
EINECSNumber (5)-0712
RTECSNumber OK9400000
TSCA Yes