missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Isobutylphenyl)-N-(1-phenylethyl)propionamide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002424
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002424 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002424 Supplier Merck Emd Millipore Supplier No. PHR2146100MG
Only null left

Specifications

CAS 105959-56-6
Linear Formula C21H27 NO
Type Certified Reference Material
Quantity 100 mg
Formula Weight 309.45
Grade Certified Reference Material