missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Hydroxyphenyl)ethanol 98.0+%, TCI America™

Catalog No. H0720500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0720500G 500 g
H072025G 25 g
H0720100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0720500G Supplier TCI America Supplier No. H0720500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 501-94-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002902
InChI Key YCCILVSKPBXVIP-UHFFFAOYSA-N
Synonym tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol
PubChem CID 10393
ChEBI CHEBI:1879
IUPAC Name 4-(2-hydroxyethyl)phenol
SMILES C1=CC(=CC=C1CCO)O

Specifications

Melting Point 91°C
Color White-Yellow
Boiling Point 310°C
Quantity 500 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Hydroxyphenyl)ethanol
EINECSNumber (3)-3710
TSCA Yes