missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Hydroxyphenyl)ethanol, >99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014349
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014349 500 mg
111014348 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014349 Supplier Merck Emd Millipore Supplier No. 79058500MGF

2-(4-Hydroxyphenyl)ethanol is a phenolic compound present in olives, which can exhibit a wide range of health benefits and pharmacological properties like anti-inflammatory, antiviral, anti-tumor and cardioprotection activity.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 501-94-0
Physical Form Neat
Linear Formula HOC6H4 CH2 CH2 OH
MDL Number MFCD00002902
Melting Point 89°C to 92°C; 90°C to 93°C
Synonym 4-Hydroxyphenethyl alcohol; Tyrosol; p-HPEA
Type Analytical Standard
Quantity 500 mg
Formula Weight 138.16
Percent Purity >99.5% (GC)
Grade Analytical Standard
Show More Show Less