missing translation for 'onlineSavingsMsg'
Learn More

2'-(4-Hydroxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5'-bibenzimidazole Trihydrochloride Hydrate 98.0+%, TCI America™

Catalog No. H134325MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H134325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H134325MG Supplier TCI America Supplier No. H134325MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

[for Biochemical Research]

Chemical Identifiers

CAS 23491-45-4
Molecular Formula C25H37Cl3N6O6
Molecular Weight (g/mol) 623.957
MDL Number MFCD00012679
InChI Key URHMSNPWJDDZSH-UHFFFAOYSA-N
Synonym bisbenzimide h 33258 fluorochrome trihydrochloride
PubChem CID 18386398
IUPAC Name 4-[5-[6-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1,3-dihydrobenzimidazol-2-ylidene]cyclohexa-2,5-dien-1-one;pentahydrate;trihydrochloride
SMILES CN1CCN(CC1)C2=CC3=C(C=C2)N=C(N3)C4=CC5=C(C=C4)NC(=C6C=CC(=O)C=C6)N5.O.O.O.O.O.Cl.Cl.Cl

Specifications

Melting Point 280°C
Quantity 25 mg
Formula Weight 533.88
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-(4-Hydroxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bibenzimidazole Trihydrochloride Hy
RTECSNumber SM1140500
TSCA Yes
Recommended Storage Freezer