missing translation for 'onlineSavingsMsg'
Learn More

2,4-Hexadiyne-1,6-diol 99.0+%, TCI America™

Catalog No. p-7130639
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06191G 1 g
H06195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06191G Supplier TCI America Supplier No. H06191G
Only null left

Chemical Identifiers

CAS 3031-68-3
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00020619
InChI Key JXMQYKBAZRDVTC-UHFFFAOYSA-N
Synonym 2,4-hexadiyne-1,6-diol, diacetylene glycol, 2,4-hexadiynediol, 2,4-hexadiyn-1,6-diol, 2,6-diol, acmc-1ahii, hoch2c=c-c=cch2oh, 1,6-dihydroxy-2,4-hexadiyne, hexa-2, 4-diyne-1, 6-diol, jxmqykbazrdvtc-uhfffaoysa
PubChem CID 94973
IUPAC Name hexa-2,4-diyne-1,6-diol
SMILES C(C#CC#CCO)O

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 1 g
UN Number 1325
Formula Weight 110.11
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Hexadiyne-1,6-diol
TSCA No
Recommended Storage Refrigerator