missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diphenyl-1,3-thiazole-5-carboxylic acid, ≥97%, Thermo Scientific™

Chemical Identifiers

CAS 502935-47-9
Molecular Formula C16H11NO2S
Molecular Weight (g/mol) 281.329
MDL Number MFCD07348756
InChI Key KMOCHRNIGWCEJV-UHFFFAOYSA-N
Synonym 2,4-diphenylthiazole-5-carboxylic acid, diphenyl-1,3-thiazole-5-carboxylic acid, 5-thiazolecarboxylicacid, 2,4-diphenyl
PubChem CID 7131193
IUPAC Name 2,4-diphenyl-1,3-thiazole-5-carboxylic acid
SMILES C1=CC=C(C=C1)C2=C(SC(=N2)C3=CC=CC=C3)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC42301CB
SDS View Documents
Thermo Scientific Chemicals
CC42301CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC42301DA
SDS View Documents
Thermo Scientific Chemicals
CC42301DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC42301EA
SDS View Documents
Thermo Scientific Chemicals
CC42301EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

502935-47-9
281.329
KMOCHRNIGWCEJV-UHFFFAOYSA-N
7131193
C1=CC=C(C=C1)C2=C(SC(=N2)C3=CC=CC=C3)C(=O)O
C16H11NO2S
MFCD07348756
2,4-diphenylthiazole-5-carboxylic acid, diphenyl-1,3-thiazole-5-carboxylic acid, 5-thiazolecarboxylicacid, 2,4-diphenyl
2,4-diphenyl-1,3-thiazole-5-carboxylic acid

Specifications

502935-47-9
100.0
Amber glass bottle
MFCD07348756
2,4-diphenylthiazole-5-carboxylic acid, diphenyl-1,3-thiazole-5-carboxylic acid, 5-thiazolecarboxylicacid, 2,4-diphenyl
C1=CC=C(C=C1)C2=C(SC(=N2)C3=CC=CC=C3)C(=O)O
281.329
281.33
2, 4-Diphenyl-1, 3-thiazole-5-carboxylic acid
96.5
97%
C16H11NO2S
250mg
KMOCHRNIGWCEJV-UHFFFAOYSA-N
2,4-diphenyl-1,3-thiazole-5-carboxylic acid
7131193
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'