missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrotoluene 99.0+%, TCI America™

Catalog No. D085625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D085625G 25 g
D0856500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D085625G Supplier TCI America Supplier No. D085625G
Only null left

Chemical Identifiers

CAS 121-14-2
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.135
MDL Number MFCD00007172
InChI Key RMBFBMJGBANMMK-UHFFFAOYSA-N
Synonym 2,4-dinitrotoluene, 2,4-dinitrotoluol, 2,4-dnt, benzene, 1-methyl-2,4-dinitro, 2,4-dinitromethylbenzene, toluene, 2,4-dinitro, rcra waste number u105, 2,4-dinitro-1-methylbenzene, 4-methyl-1,3-dinitrobenzene, dinitrotoluol
PubChem CID 8461
ChEBI CHEBI:920
IUPAC Name 1-methyl-2,4-dinitrobenzene
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 71°C
Color Yellow
Boiling Point 300°C
Quantity 25 g
UN Number 3454
Formula Weight 182.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dinitrotoluene
EINECSNumber (3)-0446
RTECSNumber XT1575000
TSCA Yes