missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrophenylacetic Acid 98.0+%, TCI America™

Catalog No. D176825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D176825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D176825G Supplier TCI America Supplier No. D176825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 643-43-6
Molecular Formula C8H6N2O6
Molecular Weight (g/mol) 226.144
MDL Number MFCD00007227
InChI Key KCNISYPADDTFDO-UHFFFAOYSA-N
Synonym 2,4-dinitrophenylacetic acid, 2-2,4-dinitrophenyl acetic acid, 2,4-dinitrophenyl acetic acid, benzeneacetic acid, 2,4-dinitro, acmc-209nlg, 2,4-dimtrophenylacetic acid, ncistruc1_000234, ncistruc2_000110, 2,4-dinitrobenzeneacetic acid, benzeneacetic acid,4-dinitro
PubChem CID 221513
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])CC(=O)O

Specifications

Melting Point 189°C
Color White-Yellow
Quantity 25 g
UN Number 1479
Formula Weight 226.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dinitrophenylacetic Acid
RTECSNumber AH2230000
TSCA Yes