missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrobromobenzene 98.0+%, TCI America™

Catalog No. D270125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D270125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D270125G Supplier TCI America Supplier No. D270125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 584-48-5
Molecular Formula C6H3BrN2O4
Molecular Weight (g/mol) 247.004
MDL Number MFCD00041873
InChI Key PBOPJYORIDJAFE-UHFFFAOYSA-N
Synonym 2,4-dinitrobromobenzene, 4-bromo-1,3-dinitrobenzene, o,p-dinitrophenyl bromide, 2,4-dinitrophenyl bromide, dnbb, benzene, 1-bromo-2,4-dinitro, 2, 4-dinitrobromobenzene, 1-bromo-2,4-dinitrobenze, unii-4ms3ftw380, ccris 1801
PubChem CID 11441
ChEBI CHEBI:58999
IUPAC Name 1-bromo-2,4-dinitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])Br

Specifications

Melting Point 72°C
Color Yellow
Quantity 25 g
UN Number 2811
Formula Weight 247.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dinitrobromobenzene
RTECSNumber CY9021500
TSCA Yes