missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-3-furoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 15058-72-7
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
InChI Key JFXXYKVSLHRBAA-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-furoic acid, 2,4-dimethyl-3-furancarboxylic acid, 3-furancarboxylic acid, 2,4-dimethyl, 2,4-dimethyl-furan-3-carboxylic acid, 2,4-dimethylfurane-3-carboxylic acid
PubChem CID 638456
IUPAC Name 2,4-dimethylfuran-3-carboxylic acid
SMILES CC1=COC(=C1C(=O)O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC65801CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC65801DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC65801DE
View Documents Promotion Details
N/A 5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

15058-72-7
140.138
2,4-dimethyl-3-furoic acid, 2,4-dimethyl-3-furancarboxylic acid, 3-furancarboxylic acid, 2,4-dimethyl, 2,4-dimethyl-furan-3-carboxylic acid, 2,4-dimethylfurane-3-carboxylic acid
2,4-dimethylfuran-3-carboxylic acid
C7H8O3
JFXXYKVSLHRBAA-UHFFFAOYSA-N
638456
CC1=COC(=C1C(=O)O)C

Specifications

15058-72-7
C7H8O3
JFXXYKVSLHRBAA-UHFFFAOYSA-N
2,4-dimethylfuran-3-carboxylic acid
638456
97%
97%
2,4-dimethyl-3-furoic acid, 2,4-dimethyl-3-furancarboxylic acid, 3-furancarboxylic acid, 2,4-dimethyl, 2,4-dimethyl-furan-3-carboxylic acid, 2,4-dimethylfurane-3-carboxylic acid
CC1=COC(=C1C(=O)O)C
140.138
140.14
2, 4-Dimethyl-3-furoic acid
Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents