missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. D41065G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D41065G Supplier TCI America Supplier No. D41065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6496-89-5
Molecular Formula C10H11O4
Molecular Weight (g/mol) 195.20
MDL Number MFCD00183328
InChI Key ZFXFMGARFHRTTO-UHFFFAOYSA-M
Synonym 2,4-dimethoxyphenylacetic acid, 2-2,4-dimethoxyphenyl acetic acid, 2,4-dimethoxyphenyl acetic acid, benzeneacetic acid, 2,4-dimethoxy, 2,4-dimethoxybenzeneacetic acid, acmc-209nop, ksc497q2d, 2,4-dimethoxyphenylaceticacid, 2, 4-dimethoxyphenylacetic acid
PubChem CID 350555
IUPAC Name 2-(2,4-dimethoxyphenyl)acetate
SMILES COC1=CC=C(CC([O-])=O)C(OC)=C1

Specifications

Melting Point 111°C
Quantity 5 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxyphenylacetic Acid