missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyphenyl isothiocyanate, 95%, Thermo Scientific™

Catalog No. AC441780010
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441780010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC441780010 Supplier Thermo Scientific Chemicals Supplier No. 441780010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33904-03-9
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.24
InChI Key CNXSCEPTSXPBTP-UHFFFAOYSA-N
Synonym 2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2,4-dimethoxy-benzene, 2,4-dimethoxybenzenisothiocyanate, acmc-20aock, 2,4-dimethoxyphenylisothiocyanate, 1-isothiocyanato-2.4-dimethoxybenzene, isothiocyanic acid 2,4-dimethoxyphenyl ester
PubChem CID 2736202
IUPAC Name 1-isothiocyanato-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)N=C=S)OC

Specifications

CAS Min % 94.0
CAS Max % 100.0
Boiling Point 166.0°C to 167.0°C (6.0mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3O)2C6H3NCS
Quantity 1g
Formula Weight 195.24
Percent Purity 95%
Chemical Name or Material 2, 4-Dimethoxyphenyl isothiocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only