missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethoxybenzylamine 98.0+%, TCI America™
$89.48 - $255.38
Chemical Identifiers
| CAS | 20781-20-8 |
|---|---|
| Molecular Formula | C9H13NO2 |
| Molecular Weight (g/mol) | 167.208 |
| MDL Number | MFCD00052393 |
| InChI Key | QOWBXWFYRXSBAS-UHFFFAOYSA-N |
| Synonym | 2,4-dimethoxybenzylamine, 2,4-dimethoxyphenyl methanamine, 2,4-dimethoxy benzylamine, 1-2,4-dimethoxyphenyl methanamine, benzenemethanamine, 2,4-dimethoxy, 2,4-dimethoxybenzyl amine, 2,4-dimethoxyphenyl methylamine, 2, 4-dimethoxybenzylamine, pubchem7398, 2,4dimethoxybenzylamine |
| PubChem CID | 597250 |
| IUPAC Name | (2,4-dimethoxyphenyl)methanamine |
| SMILES | COC1=CC(=C(C=C1)CN)OC |
Chemical Identifiers
| 20781-20-8 | |
| 167.208 | |
| QOWBXWFYRXSBAS-UHFFFAOYSA-N | |
| 597250 | |
| COC1=CC(=C(C=C1)CN)OC |
| C9H13NO2 | |
| MFCD00052393 | |
| 2,4-dimethoxybenzylamine, 2,4-dimethoxyphenyl methanamine, 2,4-dimethoxy benzylamine, 1-2,4-dimethoxyphenyl methanamine, benzenemethanamine, 2,4-dimethoxy, 2,4-dimethoxybenzyl amine, 2,4-dimethoxyphenyl methylamine, 2, 4-dimethoxybenzylamine, pubchem7398, 2,4dimethoxybenzylamine | |
| (2,4-dimethoxyphenyl)methanamine |
Specifications
| 20781-20-8 | |
| 143°C | |
| MFCD00052393 | |
| 2735 | |
| QOWBXWFYRXSBAS-UHFFFAOYSA-N | |
| (2,4-dimethoxyphenyl)methanamine | |
| 597250 | |
| ≥98.0% (T) | |
| 2,4-Dimethoxybenzylamine |
| Yellow | |
| C9H13NO2 | |
| 5 g | |
| 2,4-dimethoxybenzylamine, 2,4-dimethoxyphenyl methanamine, 2,4-dimethoxy benzylamine, 1-2,4-dimethoxyphenyl methanamine, benzenemethanamine, 2,4-dimethoxy, 2,4-dimethoxybenzyl amine, 2,4-dimethoxyphenyl methylamine, 2, 4-dimethoxybenzylamine, pubchem7398, 2,4dimethoxybenzylamine | |
| COC1=CC(=C(C=C1)CN)OC | |
| 167.208 | |
| 167.21 | |
| Liquid |
Safety and Handling
TSCA : No