missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethoxybenzylamine 98.0+%, TCI America™
Supplier: TCI America D34945G
Specifications
| 2,4-Dimethoxybenzylamine | |
| Yellow | |
| C9H13NO2 | |
| 5 g | |
| 2,4-dimethoxybenzylamine, 2,4-dimethoxyphenyl methanamine, 2,4-dimethoxy benzylamine, 1-2,4-dimethoxyphenyl methanamine, benzenemethanamine, 2,4-dimethoxy, 2,4-dimethoxybenzyl amine, 2,4-dimethoxyphenyl methylamine, 2, 4-dimethoxybenzylamine, pubchem7398, 2,4dimethoxybenzylamine | |
| COC1=CC(=C(C=C1)CN)OC | |
| 167.208 | |
| 167.21 | |
| Liquid |
| 20781-20-8 | |
| 143°C | |
| MFCD00052393 | |
| 2735 | |
| QOWBXWFYRXSBAS-UHFFFAOYSA-N | |
| (2,4-dimethoxyphenyl)methanamine | |
| 597250 | |
| ≥98.0% (T) |
Chemical Identifiers
| 20781-20-8 | |
| 167.208 | |
| QOWBXWFYRXSBAS-UHFFFAOYSA-N | |
| 597250 | |
| COC1=CC(=C(C=C1)CN)OC |
| C9H13NO2 | |
| MFCD00052393 | |
| 2,4-dimethoxybenzylamine, 2,4-dimethoxyphenyl methanamine, 2,4-dimethoxy benzylamine, 1-2,4-dimethoxyphenyl methanamine, benzenemethanamine, 2,4-dimethoxy, 2,4-dimethoxybenzyl amine, 2,4-dimethoxyphenyl methylamine, 2, 4-dimethoxybenzylamine, pubchem7398, 2,4dimethoxybenzylamine | |
| (2,4-dimethoxyphenyl)methanamine |
Safety and Handling
TSCA : No