accessibility menu, dialog, popup

UserName

CAS RN 20781-20-8

IUPAC Name: 1-(2,4-dimethoxyphenyl)methanamine
Synonyms: 1-(2,4-dimethoxyphenyl)methanamine
Molecular Weight (g/mol): 167.21
Molecular Formula: C9H13NO2
InChi Key: QOWBXWFYRXSBAS-UHFFFAOYSA-N
SMILES: COC1=CC=C(CN)C(OC)=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)