missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzoic acid, 98%

Catalog No. AAA1625222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1625222 100 g
AAA1625214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1625222 Supplier Thermo Scientific Chemicals Supplier No. A1625222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91-52-1 | C9H10O4 | 182.175 g/mol

Yttrium and heavy lanthanide complexes is synthesised with 2, 4-dimethoxybenzoic acid and thermal and spectral features is studied. In the case of the electron-rich aromatic system 2,4-dimethoxybenzoic acid reduction of the intermediate HOBt ester is slow, and good.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Yttrium and heavy lanthanide complexes is synthesised with 2, 4-dimethoxybenzoic acid and thermal and spectral features is studied. In the case of the electron-rich aromatic system 2,4-dimethoxybenzoic acid reduction of the intermediate HOBt ester is slow, and good.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 91-52-1
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00002434
InChI Key GPVDHNVGGIAOQT-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dimethoxy, 2,4-dimethoxybenzoicacid, 2,4-dimethoxy benzoic acid, 2,4-dimethoxybenzoic, pubchem4949, rarechem al bo 0028, 2,4-dimethoxy-benzoicaci, 2,4 dimethoxybenzoic acid, 2.4-dimethoxybenzoic acid, dimethoxybenzoic-2,4 acid
PubChem CID 7052
IUPAC Name 2,4-dimethoxybenzoic acid
SMILES COC1=CC(=C(C=C1)C(=O)O)OC

Specifications

Melting Point 107°C to 111°C
Odor Odorless
Quantity 100 g
Beilstein 2210860
Solubility Information Slightly soluble in water.
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material 2,4-Dimethoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-074-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only