missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethoxyaniline 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Formula Weight | 153.18 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 2,4-Dimethoxyaniline |
Safety and Handling
| EINECSNumber | (3)-0616 |
| RTECSNumber | BX4200000 |
| TSCA | Yes |