missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dimethoxyacetoacetanilide 95.0+%, TCI America™

Catalog No. D2083500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2083500G 500 g
D208325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2083500G Supplier TCI America Supplier No. D2083500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16715-79-0
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00043929
InChI Key IQWUCASGTZCNKK-UHFFFAOYSA-N
PubChem CID 85564
IUPAC Name N-(2,4-dimethoxyphenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=C(C=C(C=C1)OC)OC

Specifications

Melting Point 96°C
Quantity 500 g
Formula Weight 237.26
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2′,4′-Dimethoxyacetoacetanilide
EINECSNumber (3)-0740
TSCA Yes