missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxyquinoline 97.0+%, TCI America™

Catalog No. D1753500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1753500G 500 g
D175325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1753500G Supplier TCI America Supplier No. D1753500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86-95-3
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00006744
InChI Key HDHQZCHIXUUSMK-UHFFFAOYSA-N
Synonym 2,4-quinolinediol, 2,4-dihydroxyquinoline, 4-hydroxyquinolin-2 1h-one, quinoline-2,4-diol, 4-hydroxy-2-quinolone, 4-hydroxycarbostyril, 2 1h-quinolinone, 4-hydroxy, 4-hydroxy-2-quinolinone, hydroxycarbostyril, 2-hydroxyquinolin-4 1h-one
PubChem CID 54680871
ChEBI CHEBI:75926
IUPAC Name 4-hydroxy-1H-quinolin-2-one
SMILES C1=CC=C2C(=C1)C(=CC(=O)N2)O

Specifications

Quantity 500 g
Formula Weight 161.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dihydroxyquinoline
EINECSNumber (5)-0809
RTECSNumber FG7300000
TSCA Yes