missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzoic acid, 97%, Thermo Scientific™

Numéro de catalogue. AC114870050
Modifier l'affichage
Click to view available options
Quantité:
5g
100g
500g
Emballage:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC114870050 5g Glass Bottle
AC114871000 100g Plastic bottle
AC114875000 500g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC114870050 Fournisseur Thermo Scientific Chemicals Code fournisseur 114870050

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 89-86-1
Formule moléculaire C7H6O4
Poids moléculaire (g/mol) 154.12
Numéro MDL MFCD00002451
Clé InChI UIAFKZKHHVMJGS-UHFFFAOYSA-N
Synonyme beta-resorcylic acid, p-hydroxysalicylic acid, 4-carboxyresorcinol, 4-hydroxysalicylic acid, benzoic acid, 2,4-dihydroxy, beta-resorcinolic acid, 2,4-dhba, resorcylic acid, beta, b-resorcylic acid, 2,4-dihydroxy-benzoic acid
PubChem CID 1491
Nom de l’IUPAC 2,4-dihydroxybenzoic acid
SOURIRES C1=CC(=C(C=C1O)O)C(=O)O

Spécifications

Nom ou substance chimique 2, 4-Dihydroxybenzoic acid
Nom Remarque 97%
CAS % minimum 96.0
CAS % maximal 100.0
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 97%
Emballage Glass Bottle
Formule linéaire (HO)2C6H3CO2H
Quantité 5g
Beilstein 10, 377
Indice de Merck 15, 8278
Renseignements sur la solubilité Solubility in water: 8g/L (20°C). Other solubilities: soluble in alcohol and ether,practically insoluble in benzene
Poids de formule 154.12
Pourcentage de pureté 97%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-946-9
RTECSNumber VH3708050
TSCA TSCA

RUO – Research Use Only