missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoropyridine 98.0+%, TCI America™

Catalog No. D41735G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41735G 5 g
D41731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41735G Supplier TCI America Supplier No. D41735G
Only null left

Chemical Identifiers

CAS 34941-90-7
Molecular Formula C5H3F2N
Molecular Weight (g/mol) 115.083
MDL Number MFCD04114162
InChI Key WLAKUAILRGATSL-UHFFFAOYSA-N
Synonym 2,4-difluoro-pyridine, pyridine, 2,4-difluoro, 2,4-bis fluoranyl pyridine, 2,4-diffluoropyridine, pubchem16929, acmc-209ibr, ksc221s7f, abbypharma ap-14-5382
PubChem CID 2762838
IUPAC Name 2,4-difluoropyridine
SMILES C1=CN=C(C=C1F)F

Specifications

Color Yellow
Boiling Point 106°C
Quantity 5 g
UN Number 1993
Formula Weight 115.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Difluoropyridine