missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Difluorobenzonitrile 98.0+%, TCI America™
Supplier: TCI America D182625G
Specifications
| 2,4-Difluorobenzonitrile | |
| 47°C | |
| 83°C | |
| MFCD00009826 | |
| 2,4-difluorobenzenecarbonitrile, benzonitrile, 2,4-difluoro, 2,4-difluoro-benzonitrile, pubchem1563, acmc-1bohp, 2.4-difluoro benzonitrile, 2,4-difluoro benzonitrile, ksc493m5d, 2,4-difluorobenzonitrile, ljfdxxukkmeqke-uhfffaoysa | |
| C1=CC(=C(C=C1F)F)C#N | |
| 139.11 | |
| 139.10 | |
| Crystalline Powder |
| 3939-09-1 | |
| White-Yellow | |
| C7H3F2N | |
| 25 g | |
| LJFDXXUKKMEQKE-UHFFFAOYSA-N | |
| 2,4-difluorobenzonitrile | |
| 77545 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3939-09-1 | |
| 139.11 | |
| LJFDXXUKKMEQKE-UHFFFAOYSA-N | |
| 77545 | |
| C1=CC(=C(C=C1F)F)C#N |
| C7H3F2N | |
| MFCD00009826 | |
| 2,4-difluorobenzenecarbonitrile, benzonitrile, 2,4-difluoro, 2,4-difluoro-benzonitrile, pubchem1563, acmc-1bohp, 2.4-difluoro benzonitrile, 2,4-difluoro benzonitrile, ksc493m5d, 2,4-difluorobenzonitrile, ljfdxxukkmeqke-uhfffaoysa | |
| 2,4-difluorobenzonitrile |
Safety and Handling
TSCA : No