accessibility menu, dialog, popup

UserName

FNF

CAS RN 3939-09-1

IUPAC Name: 2,4-difluorobenzonitrile
Synonyms: 2,4-difluorobenzonitrile
Molecular Weight (g/mol): 139.11
Molecular Formula: C7H3F2N
InChi Key: LJFDXXUKKMEQKE-UHFFFAOYSA-N
SMILES: FC1=CC=C(C#N)C(F)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)