missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzonitrile 98.0+%, TCI America™

Catalog No. p-7126952
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18265G 5 g
D182625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18265G Supplier TCI America Supplier No. D18265G
Only null left

Chemical Identifiers

CAS 3939-09-1
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.11
MDL Number MFCD00009826
InChI Key LJFDXXUKKMEQKE-UHFFFAOYSA-N
Synonym 2,4-difluorobenzenecarbonitrile, benzonitrile, 2,4-difluoro, 2,4-difluoro-benzonitrile, pubchem1563, acmc-1bohp, 2.4-difluoro benzonitrile, 2,4-difluoro benzonitrile, ksc493m5d, 2,4-difluorobenzonitrile, ljfdxxukkmeqke-uhfffaoysa
PubChem CID 77545
IUPAC Name 2,4-difluorobenzonitrile
SMILES C1=CC(=C(C=C1F)F)C#N

Specifications

Melting Point 47°C
Color White-Yellow
Boiling Point 83°C
Quantity 5 g
Formula Weight 139.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Difluorobenzonitrile