missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzoic acid, 98%

CAS: 1583-58-0 | C7H4F2O2 | 158.10 g/mol

$73.42 - $245.38

Chemical Identifiers

CAS 1583-58-0
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00011670
InChI Key NJYBIFYEWYWYAN-UHFFFAOYSA-N
Synonym 2,4-dfba, benzoic acid, 2,4-difluoro, 2,4-difluorobenzoicacid, 2,4-difluoro-benzoic acid, zlchem 472, pubchem1336, acmc-1brgf, 2,4-difluorbenzoic acid, 2,4-difluorobezoic acid, 2 4-difluorobenzoic acid
PubChem CID 74102
IUPAC Name 2,4-difluorobenzoic acid
SMILES OC(=O)C1=CC=C(F)C=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1340306
View Documents
Thermo Scientific Chemicals
A1340306
5 g N/A
Only null left
 
AAA1340314
View Documents
Thermo Scientific Chemicals
A1340314
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1583-58-0
158.10
NJYBIFYEWYWYAN-UHFFFAOYSA-N
74102
OC(=O)C1=CC=C(F)C=C1F
C7H4F2O2
MFCD00011670
2,4-dfba, benzoic acid, 2,4-difluoro, 2,4-difluorobenzoicacid, 2,4-difluoro-benzoic acid, zlchem 472, pubchem1336, acmc-1brgf, 2,4-difluorbenzoic acid, 2,4-difluorobezoic acid, 2 4-difluorobenzoic acid
2,4-difluorobenzoic acid

Specifications

1583-58-0
Odorless
MFCD00011670
973355
NJYBIFYEWYWYAN-UHFFFAOYSA-N
2,4-difluorobenzoic acid
74102
98%
186°C to 190°C
C7H4F2O2
5 g
2,4-dfba, benzoic acid, 2,4-difluoro, 2,4-difluorobenzoicacid, 2,4-difluoro-benzoic acid, zlchem 472, pubchem1336, acmc-1brgf, 2,4-difluorbenzoic acid, 2,4-difluorobezoic acid, 2 4-difluorobenzoic acid
OC(=O)C1=CC=C(F)C=C1F
158.10
158.11
2,4-Difluorobenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 216-430-9

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only