Learn More
2,4-Difluorobenzoic acid, 98%
CAS: 1583-58-0 | C7H4F2O2 | 158.10 g/mol
Supplier: Thermo Scientific Chemicals A1340306
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,4-Difluorobenzoic acid | |
| 186°C to 190°C | |
| C7H4F2O2 | |
| 5 g | |
| 2,4-dfba, benzoic acid, 2,4-difluoro, 2,4-difluorobenzoicacid, 2,4-difluoro-benzoic acid, zlchem 472, pubchem1336, acmc-1brgf, 2,4-difluorbenzoic acid, 2,4-difluorobezoic acid, 2 4-difluorobenzoic acid | |
| OC(=O)C1=CC=C(F)C=C1F | |
| 158.10 | |
| 158.11 |
| 1583-58-0 | |
| Odorless | |
| MFCD00011670 | |
| 973355 | |
| NJYBIFYEWYWYAN-UHFFFAOYSA-N | |
| 2,4-difluorobenzoic acid | |
| 74102 | |
| 98% |
Chemical Identifiers
| 1583-58-0 | |
| 158.10 | |
| NJYBIFYEWYWYAN-UHFFFAOYSA-N | |
| 74102 | |
| OC(=O)C1=CC=C(F)C=C1F |
| C7H4F2O2 | |
| MFCD00011670 | |
| 2,4-dfba, benzoic acid, 2,4-difluoro, 2,4-difluorobenzoicacid, 2,4-difluoro-benzoic acid, zlchem 472, pubchem1336, acmc-1brgf, 2,4-difluorbenzoic acid, 2,4-difluorobezoic acid, 2 4-difluorobenzoic acid | |
| 2,4-difluorobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 216-430-9
TSCA : Yes
RUO – Research Use Only