Learn More
2,4-Difluoroaniline, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 114590100
| Quantity | 10mL |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C6H5F2N | |
| MFCD00007648 | |
| 2,4-difluorobenzenamine, benzenamine, 2,4-difluoro, 2,4-difluroaniline, 2,4-difluorophenylamine, aniline, 2,4-difluoro, unii-40p93l7kwd, 1-amino-2,4-difluorobenzene, ccris 4621, 2,4-difluoro aniline, 2,4-difluoro-phenylamine | |
| 2,4-difluoroaniline |
Specifications
| 2, 4-Difluoroaniline | |
| 98.5 | |
| 1.2680g/mL | |
| 62°C | |
| 98.5% min. (GC) | |
| C6H5F2N | |
| F2C6H3NH2 | |
| MFCD00007648 | |
| 1.268 | |
| 2,4-difluorobenzenamine, benzenamine, 2,4-difluoro, 2,4-difluroaniline, 2,4-difluorophenylamine, aniline, 2,4-difluoro, unii-40p93l7kwd, 1-amino-2,4-difluorobenzene, ccris 4621, 2,4-difluoro aniline, 2,4-difluoro-phenylamine | |
| CEPCPXLLFXPZGW-UHFFFAOYSA-N | |
| 2,4-difluoroaniline | |
| 9709 | |
| 99% |
| 367-25-9 | |
| 100.0 | |
| 170°C (753.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.5025 to 1.5075 | |
| 10mL | |
| 12, I, 297 | |
| 15, 3166 | |
| Solubility in water: immiscible | |
| NC1=CC=C(F)C=C1F | |
| 129.11 | |
| 129.11 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfort
GHS Signal Word: Danger
EINECSNumber : 206-687-5
Recommended Storage : Refrigerator +4°C