missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-3-nitrobenzonitrile, 97%, Thermo Scientific Chemicals

Catalog No. AC451420010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451420010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC451420010 Supplier Thermo Scientific Chemicals Supplier No. 451420010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1186194-75-1 | Molecular Formula: C7H2F2N2O2 | Molecular Weight: 184.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1186194-75-1
Molecular Formula C7H2F2N2O2
Molecular Weight (g/mol) 184.1
InChI Key HESWWHRCQMLPFT-UHFFFAOYSA-N
Synonym 3-nitro-2,4-difluoro-benzonitrile
PubChem CID 45790497
IUPAC Name 2,4-difluoro-3-nitrobenzonitrile
SMILES C1=CC(=C(C(=C1C#N)F)[N+](=O)[O-])F

Specifications

Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Refractive Index 1.529
Quantity 1 g
Formula Weight 184.1
Percent Purity 97%
Chemical Name or Material 2, 4-Difluoro-3-nitrobenzonitrile
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of

RUO – Research Use Only