missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-3-methoxyaniline, 97%

Catalog No. AAH2648906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2648906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2648906 Supplier Thermo Scientific Chemicals Supplier No. H2648906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 886499-08-7 | C7H7F2NO | 159.14 g/mol

2,4-Difluoro-3-methoxyaniline is a fluorinated compound used as a building block in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoro-3-methoxyaniline is a fluorinated compound used as a building block in organic synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 886499-08-7
Molecular Formula C7H7F2NO
Molecular Weight (g/mol) 159.14
MDL Number MFCD04115929
InChI Key ISMLABQIDHXORP-UHFFFAOYSA-N
Synonym 2,4-difluoro-3-methoxyphenylamine, 2,4-difluoro-3-methoxy-aniline, benzenamine, 2,4-difluoro-3-methoxy
PubChem CID 3865162
IUPAC Name 2,4-difluoro-3-methoxyaniline
SMILES COC1=C(F)C=CC(N)=C1F

Specifications

Refractive Index 1.513
Quantity 5 g
UN Number UN2810
Solubility Information Immiscible with water.
Formula Weight 159.14
Percent Purity 97%
Chemical Name or Material 2,4-Difluoro-3-methoxyaniline
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only