missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diethylthioxanthen-9-one 98.0+%, TCI America™

Catalog No. D237525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D237525G 25 g
D2375250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D237525G Supplier TCI America Supplier No. D237525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82799-44-8
Molecular Formula C17H16OS
Molecular Weight (g/mol) 268.374
MDL Number MFCD00082910
InChI Key BTJPUDCSZVCXFQ-UHFFFAOYSA-N
PubChem CID 122825
IUPAC Name 2,4-diethylthioxanthen-9-one
SMILES CCC1=CC(=C2C(=C1)C(=O)C3=CC=CC=C3S2)CC

Specifications

Melting Point 70°C
Color Yellow
Quantity 25 g
Formula Weight 268.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diethylthioxanthen-9-one
EINECSNumber (5)-5645
TSCA Yes