missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzyl chloride, 98%

Catalog No. AAA1143536
Change view
Click to view available options
Quantity:
500 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1143536 500 g
AAA114350B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1143536 Supplier Thermo Scientific Chemicals Supplier No. A1143536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 94-99-5 | C7H5Cl3 | 195.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94-99-5
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.47
MDL Number MFCD00000895
InChI Key IRSVDHPYXFLLDS-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzyl chloride, 2,4-dichloro-1-chloromethyl benzene, 2,4-dichlorobenzylchloride, alpha,2,4-trichlorotoluene, benzene, 2,4-dichloro-1-chloromethyl, 2,4-dichloro-1-chloromethyl-benzene, alpha2,4-trichlorotoluene, unii-85e96p2s3s, 2,4-dcbc, toluene, .alpha.2,4-trichloro
PubChem CID 7212
IUPAC Name 2,4-dichloro-1-(chloromethyl)benzene
SMILES ClCC1=CC=C(Cl)C=C1Cl

Specifications

Melting Point -3°C
Density 1.408
Boiling Point 248°C
Flash Point 190°C (374°F)
Odor Pungent
Refractive Index 1.576
Quantity 500 g
UN Number UN3265
Beilstein 387220
Formula Weight 195.48
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorobenzyl chloride
Show More Show Less
Hazard Category H302-H314-H332-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 202-381-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only