missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzaldehyde, 98%

Catalog No. AC164140050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164140050 5 g Glass bottle
AC164141000 100 g Plastic bottle
AC164145000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164140050 Supplier Thermo Scientific Chemicals Supplier No. 164140050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 874-42-0 | C7H4Cl2O | 175.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 874-42-0
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00003305
InChI Key YSFBEAASFUWWHU-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dichloro, unii-z08ql2124r, ccris 6013, 2,4-dichloro benzaldehyde, 2,4-dichloro-benzaldehyde, acmc-209qnb, dsstox_cid_2130, 2,4 dichloro benzaldehyde, 2,4-di-chlorobenzaldehyde, dsstox_rid_76500
PubChem CID 13404
IUPAC Name 2,4-dichlorobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)Cl)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 69°C to 73°C
Color Yellow to Beige
Boiling Point 233°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3CHO
Quantity 5 g
Beilstein 07, 236
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in methanol,ethanol,ether and acetone
Formula Weight 175.01
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2, 4-Dichlorobenzaldehyde, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-861-1
TSCA TSCA

RUO – Research Use Only