missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylaniline, 97%

Catalog No. AAA1259122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1259122 100 g
AAA1259106 5 g
AAA1259114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1259122 Supplier Thermo Scientific Chemicals Supplier No. A1259122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30273-00-8 | C7H7Cl2N | 176.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30273-00-8
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00044074
InChI Key UAISVUGQLKXPFF-UHFFFAOYSA-N
Synonym 4,6-dichloro-2-methylaniline, benzenamine, 2,4-dichloro-6-methyl, acmc-20aodo, 4,6-dichloro-o-toluidine, 2-amino-3,5-dichlorotoluene, 2,4-dichloro-6-methylphenylamine, 4,6-dichloro-2-methylphenylamine, 2,4-bis chloranyl-6-methyl-aniline, 2,4-dichloro-6-methylaniline
PubChem CID 1268287
IUPAC Name 2,4-dichloro-6-methylaniline
SMILES CC1=CC(=CC(=C1N)Cl)Cl

Specifications

Melting Point 43°C to 45°C
Boiling Point 82°C to 84°C (0.1 mmHg)
Quantity 100 g
Beilstein 2208221
Formula Weight 176.05
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-6-methylaniline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only