missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromotoluene 98.0+%, TCI America™

Catalog No. D35915G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35915G 5 g
D35911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35915G Supplier TCI America Supplier No. D35915G

Chemical Identifiers

CAS 31543-75-6
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00052985
InChI Key GHWYNNFPUGEYEM-UHFFFAOYSA-N
Synonym 2,4-dibromotoluene, benzene, 2,4-dibromo-1-methyl, 2,4-bromotoluene, 2,4-dibromo-toluene, pubchem3915, 2,4-dibromo toluene, acmc-1bn9b, ksc497e7f, 2,4-dibromo-1-methyl-benzene, ghwynnfpugeyem-uhfffaoysa
PubChem CID 94421
IUPAC Name 2,4-dibromo-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Br)Br

Specifications

Melting Point -10°C
Color Yellow
Boiling Point 243°C
Quantity 5 g
Formula Weight 249.93
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dibromotoluene