missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromophenyl Glycidyl Ether 90.0+%, TCI America™

Numéro de catalogue. D488525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D488525G 25 g
D4885500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D488525G Fournisseur TCI America Code fournisseur D488525G
Il en reste null

Identifiants chimiques

CAS 20217-01-0
Molecular Formula C9H8Br2O2
Molecular Weight (g/mol) 307.97
MDL Number MFCD13194826
InChI Key NFWLWLQSZIJYFR-UHFFFAOYNA-N
PubChem CID 62740
IUPAC Name 2-[(2,4-dibromophenoxy)methyl]oxirane
SMILES BrC1=CC=C(OCC2CO2)C(Br)=C1

Spécifications

Melting Point 41°C
Color White-Yellow
Quantity 25 g
Formula Weight 307.97
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Dibromophenyl Glycidyl Ether
EINECSNumber (3)-0987&(3)-0921
TSCA Yes