missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromophenol 98.0+%, TCI America™

Catalog No. D0265500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0265500G 500 g
D026525G 25 g
D0265100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0265500G Supplier TCI America Supplier No. D0265500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 615-58-7
Molecular Formula C6H4Br2O
Molecular Weight (g/mol) 251.905
MDL Number MFCD00002149
InChI Key FAXWFCTVSHEODL-UHFFFAOYSA-N
Synonym phenol, 2,4-dibromo, 2,4-dibromo-phenol, 2,4-dibromphenol, unii-ia75t5c9tg, 2,4-bis bromanyl phenol, ia75t5c9tg, phenol,4-dibromo, emery 9331, acmc-1bga9, 2,4-dibromophenol
PubChem CID 12005
ChEBI CHEBI:34238
IUPAC Name 2,4-dibromophenol
SMILES C1=CC(=C(C=C1Br)Br)O

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 239°C
Quantity 500 g
UN Number 2811
Formula Weight 251.91
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dibromophenol
EINECSNumber (3)-0907
RTECSNumber SK8010000
TSCA Yes