missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromophenol, 98%

Catalog No. AAB2339422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2339422 100 g
AAB2339414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2339422 Supplier Thermo Scientific Chemicals Supplier No. B2339422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 615-58-7 | C6H4Br2O | 251.905 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 615-58-7
Molecular Formula C6H4Br2O
Molecular Weight (g/mol) 251.905
MDL Number MFCD00002149
InChI Key FAXWFCTVSHEODL-UHFFFAOYSA-N
Synonym phenol, 2,4-dibromo, 2,4-dibromo-phenol, 2,4-dibromphenol, unii-ia75t5c9tg, 2,4-bis bromanyl phenol, ia75t5c9tg, phenol,4-dibromo, emery 9331, acmc-1bga9, 2,4-dibromophenol
PubChem CID 12005
ChEBI CHEBI:34238
IUPAC Name 2,4-dibromophenol
SMILES C1=CC(=C(C=C1Br)Br)O

Specifications

Melting Point 35°C to 38°C
Boiling Point 239°C
Flash Point >110°C (230°F)
Odor Phenol-like
Quantity 100 g
UN Number UN2811
Beilstein 1861291
Formula Weight 251.92
Percent Purity 98%
Chemical Name or Material 2,4-Dibromophenol
Hazard Category H300-H315-H319-H335
Hazard Statement GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 210-436-5
RTECSNumber SK8010000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only