missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromobenzenesulfonyl chloride, 98%

Catalog No. AAL1988706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1988706 5 g
AAL1988703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1988706 Supplier Thermo Scientific Chemicals Supplier No. L1988706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 72256-95-2 | C6H3Br2ClO2S | 334.41 g/mol

2,4-Dibromobenzenesulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dibromobenzenesulfonyl chloride is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 72256-95-2
Molecular Formula C6H3Br2ClO2S
Molecular Weight (g/mol) 334.41
MDL Number MFCD03093759
InChI Key OMOXIKAWVFDFNX-UHFFFAOYSA-N
Synonym 2,4-dibromobenzene-1-sulfonyl chloride, 2,4-dibromobenzenesulphonyl chloride, 2,4-dibromophenyl chlorosulfone, pubchem5111, 2,4-dibromophenylsulfonyl chloride, benzenesulfonylchloride, 2,4-dibromo, 2,4-bis bromanyl benzenesulfonyl chloride, 2,4-dibromobenzenesulfonyl chloride
PubChem CID 2737366
IUPAC Name 2,4-dibromobenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=C(Br)C=C(Br)C=C1

Specifications

Melting Point 82°C to 86°C
Boiling Point 128°C (0.2 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 2694058
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 334.41
Percent Purity 98%
Chemical Name or Material 2,4-Dibromobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only