missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-3-pentanone, mixture of stereoisomers, 97%

Catalog No. AAA1668009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1668009 10 g
AAA1668018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1668009 Supplier Thermo Scientific Chemicals Supplier No. A1668009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 815-60-1 | C5H8Br2O | 243.926 g/mol

2,4-Dibromo-2,4-dimethyl-3-pentanone may be used in the synthesis of 2-dimethylamino-4-methylene-1,3-dioxolanes, via debromination using zinc-copper couple in dimethylformamide and dimethylacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dibromo-2,4-dimethyl-3-pentanone may be used in the synthesis of 2-dimethylamino-4-methylene-1,3-dioxolanes, via debromination using zinc-copper couple in dimethylformamide and dimethylacetamide.

Solubility
Slightly soluble in water(50 g/L (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, reducing agents, bases.

Chemical Identifiers

CAS 815-60-1
Molecular Formula C5H8Br2O
Molecular Weight (g/mol) 243.926
MDL Number MFCD00015719
InChI Key UOPIOAUZQKSZRO-UHFFFAOYSA-N
Synonym 2,4-dibromo-3-pentanone, 2,4-dibromo-pentan-3-one, 3-pentanone,2,4-dibromo, 2,4-bis bromanyl pentan-3-one, 2,4-dibromo-3-pentanone, mixture of stereoisomers
PubChem CID 11791395
IUPAC Name 2,4-dibromopentan-3-one
SMILES CC(C(=O)C(C)Br)Br

Specifications

Boiling Point 67°C to 82°C (10 mmHg)
Odor Pungent
Quantity 10 g
UN Number UN3265
Solubility Information Slightly soluble in water(50g/L (20°C).
Formula Weight 243.91
Percent Purity 97%
Chemical Name or Material 2,4-Dibromo-3-pentanone, mixture of stereoisomers
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only