missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diaminotoluene 98.0+%, TCI America™

Catalog No. D012325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D012325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D012325G Supplier TCI America Supplier No. D012325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-80-7
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00007804
InChI Key VOZKAJLKRJDJLL-UHFFFAOYSA-N
Synonym 2,4-diaminotoluene, 2,4-toluenediamine, developer t, toluene-2,4-diamine, 1,3-benzenediamine, 4-methyl, developer mtd, m-toluenediamine, fourrine m, m-tolylenediamine, benzofur mt
PubChem CID 7261
ChEBI CHEBI:34237
IUPAC Name 4-methylbenzene-1,3-diamine
SMILES CC1=CC=C(N)C=C1N

Specifications

Melting Point 98°C
Boiling Point 292°C
Quantity 25 g
UN Number 1709
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diaminotoluene
EINECSNumber (3)-0126
RTECSNumber XS9625000
TSCA Yes