missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)ethylamine 98.0+%, TCI America™

Catalog No. C11295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11295G 5 g
C112925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11295G Supplier TCI America Supplier No. C11295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 156-41-2
Molecular Formula C8H11ClN
Molecular Weight (g/mol) 156.63
MDL Number MFCD00008191
InChI Key SRXFXCKTIGELTI-UHFFFAOYSA-O
Synonym 4-chlorophenethylamine, 2-4-chlorophenyl ethylamine, 2-4-chlorophenyl ethanamine, 2-p-chlorophenyl ethylamine, p-chlorophenethylamine, 2-4-chlorophenyl ethan-1-amine, 4-chlorobenzeneethanamine, 2-4-chloro-phenyl-ethylamine, benzeneethanamine, 4-chloro, beta-p-chlorophenyl ethylamine
PubChem CID 67430
IUPAC Name 2-(4-chlorophenyl)ethan-1-aminium
SMILES [NH3+]CCC1=CC=C(Cl)C=C1

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 155.63
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(4-Chlorophenyl)ethylamine