missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)ethylamine, 97%

Catalog No. AAH6308122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6308122 100 g
AAH6308106 5 g
AAH6308114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6308122 Supplier Thermo Scientific Chemicals Supplier No. H6308122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 156-41-2 | C8H11ClN | 156.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 156-41-2
Molecular Formula C8H11ClN
Molecular Weight (g/mol) 156.63
MDL Number MFCD00008191
InChI Key SRXFXCKTIGELTI-UHFFFAOYSA-O
Synonym 4-chlorophenethylamine, 2-4-chlorophenyl ethylamine, 2-4-chlorophenyl ethanamine, 2-p-chlorophenyl ethylamine, p-chlorophenethylamine, 2-4-chlorophenyl ethan-1-amine, 4-chlorobenzeneethanamine, 2-4-chloro-phenyl-ethylamine, benzeneethanamine, 4-chloro, beta-p-chlorophenyl ethylamine
PubChem CID 67430
IUPAC Name 2-(4-chlorophenyl)ethanamine
SMILES [NH3+]CCC1=CC=C(Cl)C=C1

Specifications

Density 1.120 g/mL
Boiling Point 60°C to 65°C (0.1 mmHg)
Flash Point 106°C (223°F)
Quantity 100 g
Beilstein 508247
Formula Weight 155.62
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(4-Chlorophenyl)ethylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 205-853-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only