missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)-1-phenylbenzimidazole 98.0+%, TCI America™

Numéro de catalogue. B44791G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B44791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. B44791G Fournisseur TCI America Code fournisseur B44791G
Il en reste null

Identifiants chimiques

CAS 2620-76-0
Molecular Formula C19H13BrN2
Molecular Weight (g/mol) 349.231
MDL Number MFCD09261342
InChI Key DXRLALXPCIOIDK-UHFFFAOYSA-N
Synonym 2-4-bromophenyl-1-phenyl-1h-benzoimidazole, 2-4-bromo-phenyl-1-phenyl-1h-benzoimidazole, 2-4-bromophenyl-1-phenyl-1h-benzimidazole, 2-4-bromophenyl-1-phenylbenzimidazole, 2-4-bromophenyl-1-phenyl-1h-benzo d imidazole, 1h-benzimidazole, 2-4-bromophenyl-1-phenyl, 1-phenyl-2-4-bromophenyl benzimidazole, 2-4-bromophenyl-1-phenyl-1h-1,3-benzodiazole, 2-4-bromophenyl-1-phenyl-1,3-benzodiazole, zlchem 100
PubChem CID 23094070
IUPAC Name 2-(4-bromophenyl)-1-phenylbenzimidazole
SMILES C1=CC=C(C=C1)N2C3=CC=CC=C3N=C2C4=CC=C(C=C4)Br

Spécifications

Melting Point 168°C
Quantity 1 g
Formula Weight 349.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Bromophenyl)-1-phenylbenzimidazole