missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine 95.0+%, TCI America™

Catalog No. B4587200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4587200MG 200 mg
B45871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4587200MG Supplier TCI America Supplier No. B4587200MG

Chemical Identifiers

CAS 851030-18-7
Molecular Formula C17H14ClN3O2
Molecular Weight (g/mol) 327.77
MDL Number MFCD29089341
InChI Key XKDFUXJROYAXAO-UHFFFAOYSA-N
Synonym DBT-Cl
PubChem CID 71406442
IUPAC Name 2,4-bis(benzyloxy)-6-chloro-1,3,5-triazine
SMILES ClC1=NC(OCC2=CC=CC=C2)=NC(OCC2=CC=CC=C2)=N1

Specifications

Melting Point 66°C
Color White
Quantity 200 mg
Formula Weight 327.77
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine