missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%

Catalog No. AAH3605206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3605206 5 g
AAH3605203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3605206 Supplier Thermo Scientific Chemicals Supplier No. H3605206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 42333-76-6 | C18H12N6 | 312.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42333-76-6
Molecular Formula C18H12N6
Molecular Weight (g/mol) 312.34
MDL Number MFCD23379843
InChI Key UOSYDSDIRAXTTF-UHFFFAOYSA-N
Synonym 2,4,6-tris 3-pyridyl-s-triazine, 2,4,6-tri-3-pyridinyl-1,3,5-triazine, tris pyridin-3-yl-1,3,5-triazine, 2,4,6-tris 3-pyridyl-1,3,5-triazine
PubChem CID 11472427
IUPAC Name 2,4,6-tripyridin-3-yl-1,3,5-triazine
SMILES C1=CC(=CN=C1)C1=NC(=NC(=N1)C1=CN=CC=C1)C1=CN=CC=C1

Specifications

Melting Point >300°C
Color White to Yellow
Quantity 5 g
Formula Weight 312.33
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,4,6-Tris(3-pyridyl)-1,3,5-triazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only